您想再次登录吗?
薄层色谱
薄层色谱
Thin Layer Chromatography (TLC) and High Performance Thin Layer Chromatography (HPTLC) are often referred to as Planar Chromatography. This is a modern classical separation technique, established worldwide and distinguished by flexibility, reliability and cost efficiency.
 Thin Layer Chromatography offers:
  • Enormous flexibility
  • Cheap separation method without the need for sophisticated instruments (only for TLC)
  • Multiple samples (up to 72) can be run simultaneously under identical conditions
  • Unsurpassed clarity and simultaneous visual evaluation of all samples and sample components 
  • Simplified sample preparation due to single use of the stationary phase