您想再次登录吗?
悬浮物
悬浮物
VWR offers a variety of products for you to measure water quality from the point of view of suspended materials: 
  • Rapid method with moisture analysers
  • Centrifugation or filtration methods in compliance with European norms
  • The method for removing volatile suspended solids from the mainly inorganic substances or fixed solids involves the use of both a furnace and an analytical balance.