您想再次登录吗?
高压灭菌 & 附件
高压灭菌 & 附件
Autoclaves are used to sterilise equipment and supplies by subjecting them to high pressure saturated steam.
Typical loads include laboratory glassware, equipment, instruments and of course culture media. A growing area of autoclave usage is the pre-disposal treatment and sterilisation of waste material, such as pathogenic waste before final disposal.
Offering both bench top and the larger laboratory autoclaves, with a choice of either front or top loading, they come in a variety of sizes which are available from all major suppliers as well as our own VWR Collection range.
If you need validation of your autoclaving cycle then VWR also offers a huge portfolio of data loggers as well as other essential accessories such as baskets, trays and foil.