Total immersion probe
供应商: Cowie
COWI500.050.1EA 0 CNY
COWI500.050.1
Total immersion probe
温度探头
Designed for total immersion, suitable for use in autoclaves and most corrosive liquids.
  • PRT100 Class A sensor
  • Inert
  • Pressure resistant
  • Features 1 m long PFA insulated cable
Delivery information: Supplied without plug.