2-(1,2-Benzisoxazol-3-yl)acetic acid ≥98.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAB2912-25GEA
0
CNY
TCIAB2912-25G
2-(1,2-Benzisoxazol-3-yl)acetic acid ≥98.0% (通过GC,滴定分析)
2-(1,2-Benzisoxazol-3-yl)acetic acid
分子式:
C₉H₇NO₃ 分子量: 177,1568 g/mol |
MDL编号:
MFCD02180388 CAS号: 4865-84-3 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...