4,4'-Dimethyl-trans-stilbene ≥99.0% (通过GC)
供应商: TCI EUROPE
TCIAD2708-5GEA
0
CNY
TCIAD2708-5G
4,4'-Dimethyl-trans-stilbene ≥99.0% (通过GC)
4,4'-Dimethyl-trans-stilbene
分子式:
C₁₆H₁₆ 分子量: 208,2982 g/mol |
MDL编号:
MFCD00136904 CAS号: 18869-29-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 181.0 to 184.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...