Quinizarin green ≥98.0% (以总氮计)

供应商: TCI EUROPE
Warning

TCIAQ0021-25GEA 0 CNY
TCIAQ0021-25G
Quinizarin green ≥98.0% (以总氮计)
Quinizarin green
分子式: C₂₈H₂₂N₂O₂
分子量: 418,496 g/mol
MDL编号: MFCD00001195
CAS号: 128-80-3
EINECS: 204-909-5
https://cn.vwr.com/stibo/web/std.lang.all/36/88/28993688.jpg

现在订购

产品技术规格

Appearance Gray to Dark purple to Black powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Lambda max. 508.0 to 603.0 nm (Acetone)
Lambda max.2 635.0 to 640.0 nm (Acetone)
Absorbance (E1%1cm) min. 350 (Acetone, 508.0 to 603.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR