3'-Amino-4'-methoxyacetanilide ≥98.0% (通过HPLC,滴定分析)
ABCRAB136303.0500
3'-Amino-4'-methoxyacetanilide ≥98.0% (通过HPLC,滴定分析)
3'-Amino-4'-methoxyacetanilide
分子式:
C₉H₁₂N₂O₂ 分子量: 180,2000 g/mol |
MDL编号:
MFCD00008676 CAS号: 6375-47-9 EINECS: 228-938-8 |
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...