硫代乙酰苯胺 ≥98.0% (以总氮计)
供应商: TCI EUROPE
TCIAT0188-5GEA
0
CNY
TCIAT0188-5G
硫代乙酰苯胺 ≥98.0% (以总氮计)
硫代乙酰苯胺
分子式:
C₈H₉NS 分子量: 151,2288 g/mol 沸点: 147°C |
MDL编号:
MFCD00004942 CAS号: 637-53-6 EINECS: 211-288-4 |
![]() |
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...