N-乙基咔唑 ≥99.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAE0247-25GEA 230 CNY
TCIAE0247-25G
N-乙基咔唑 ≥99.0% (通过GC)
N-乙基咔唑
分子式: C₁₄H₁₃N
分子量: 195,27 g/mol
沸点: 166°C
MDL编号: MFCD00004967
CAS号: 86-28-2
EINECS: 201-660-4
https://cn.vwr.com/stibo/web/std.lang.all/78/02/28967802.jpg

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 68.0 to 71.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR