Potassium-2-formylphenyltrifluoroborate ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAP1812-1GEA
0
CNY
TCIAP1812-1G
Potassium-2-formylphenyltrifluoroborate ≥98.0% (通过滴定分析)
Potassium-2-formylphenyltrifluoroborate
分子式:
C₇H₅BF₃KO 分子量: 212,02 g/mol |
MDL编号:
MFCD02093979 CAS号: 192863-39-1 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 219.0 to 225.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...