4-(Dibutylamino)salicylaldehyde ≥96.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAD3260-5GEA
0
CNY
TCIAD3260-5G
4-(Dibutylamino)salicylaldehyde ≥96.0% (通过GC)
4-(Dibutylamino)salicylaldehyde
分子式:
C₁₅H₂₃NO₂ 存储条件: Freezer |
CAS号:
57771-09-2 |
产品技术规格
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...