4-(甲硫基)苯硫醇 ≥97.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
4-甲基硫代硫代苯酚
, 4-甲硫基苯硫酚
TCIAM1823-5GEA
0
CNY
TCIAM1823-5G
4-(甲硫基)苯硫醇 ≥97.0% (通过滴定分析)
4-(甲硫基)苯硫醇
分子式:
C₇H₈S₂ 分子量: 156,27 g/mol 沸点: 117°C |
MDL编号:
MFCD00082770 CAS号: 1122-97-0 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Iodometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...