1-苯并噻酚-2-羧醛 ≥98.0% (通过GC)

供应商: TCI EUROPE

Synonyms: 苯并噻吩-2-甲醛

TCIAB3777-5GEA 0 CNY
TCIAB3777-5G
1-苯并噻酚-2-羧醛 ≥98.0% (通过GC)
1-苯并噻酚-2-羧醛
分子式: C₉H₆OS
分子量: 162,21 g/mol
沸点: 136°C
MDL编号: MFCD01075041
CAS号: 3541-37-5
https://cn.vwr.com/stibo/web/std.lang.all/51/13/28935113.jpg

现在订购

产品技术规格

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR