2-噻吩甲酰胺 ≥98.0% (以总氮计)
供应商: TCI EUROPE
TCIAT1252-5GEA
0
CNY
TCIAT1252-5G
2-噻吩甲酰胺 ≥98.0% (以总氮计)
2-噻吩甲酰胺
分子式:
C₅H₅NOS 分子量: 127,17 g/mol |
MDL编号:
MFCD00010425 CAS号: 5813-89-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 180.0 to 183.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...