3-乙氧基苯硫酚 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAE0701-5GEA
0
CNY
TCIAE0701-5G
TCIAE0701-1G
3-乙氧基苯硫酚 ≥98.0% (通过GC)
3-乙氧基苯硫酚
分子式:
C₈H₁₀OS 沸点: 239°C |
MDL编号:
MFCD03094660 CAS号: 86704-82-7 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...