反-1,3-戊二烯 ≥93.0% (通过GC) stabilised

供应商: TCI EUROPE
Danger

Synonyms: 反戊间二稀

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

TCIAP0647-5MLEA 0 CNY
TCIAP0647-5ML
反-1,3-戊二烯 ≥93.0% (通过GC) stabilised
反-1,3-戊二烯
分子式: C₅H₈
沸点: 42°C
闪点: -29°C
存储条件: Refrigerator
MDL编号: MFCD00009295
CAS号: 2004-70-8
EINECS: 217-909-5
UN号: 3295
ADR: 3,II
https://cn.vwr.com/stibo/web/std.lang.all/13/67/28991367.jpg

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %
Specific gravity (20/20) 0.6740 to 0.6840

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR