乙基二苯基膦 ≥97.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
乙二苯膦
TCIAE0519-5GEA
0
CNY
TCIAE0519-5G
乙基二苯基膦 ≥97.0% (通过GC)
乙基二苯基膦
分子式:
C₁₄H₁₅P 分子量: 214,24 g/mol 沸点: 157°C |
MDL编号:
MFCD00015170 CAS号: 607-01-2 ADR: 6.1,III |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...