Wash bottles, glass
供应商: Hecht
HECH43091048EA 810.78 CNY
HECH43091048
Wash bottles, glass
Universal wash bottle, made of borosilicate glass 3.3, consisting of a 250 ml or 1000 ml capacity flat bottomed flask with standard ground joint sleeve NS 29/32 and spray assembly with standard ground joint cone NS 29/32.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Fully interchangeable parts