Magnetic stirring bars, triangle with rib
供应商: Cowie
COWI001.1812EA 0 CNY
COWI001.1812
Magnetic stirring bars, triangle with rib
搅拌器 磁力搅拌子
These PTFE triangle with rib magnetic stirring bars have excellent turbulence even at low speed and can be used for slightly curved or uneven surfaces.