2-氨基-3-甲酰色酮 ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Warning

Synonyms: 2-氨基-4-氧代-4H-1-苯并吡喃-3-甲醛

TCIAA2415-1GEA 0 CNY
TCIAA2415-1G
2-氨基-3-甲酰色酮 ≥98.0% (通过HPLC,总氮)
2-氨基-3-甲酰色酮
分子式: C₁₀H₇NO₃
分子量: 189,1675 g/mol
溶点: 250°C
MDL编号: MFCD00191735
CAS号: 61424-76-8
https://cn.vwr.com/stibo/web/std.lang.all/05/99/28930599.jpg

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR