Cholesterol isopropyl carbonate ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAC0717-10GEA 0 CNY
TCIAC0717-10G
Cholesterol isopropyl carbonate ≥95.0% (通过GC)
Cholesterol isopropyl carbonate
分子式: C₃₁H₅₂O₃
MDL编号: MFCD00059507
CAS号: 78916-25-3
https://cn.vwr.com/stibo/web/std.lang.all/77/63/28937763.jpg

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Mesomorphic range 105.0 to 111.0 deg-C
Specific rotation [a]20/D -24.0 to -30.0 deg (C=5, THF)
Solubility in THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR