Cholesterol octyl carbonate

供应商: TCI EUROPE

TCIAC0721-10GEA 0 CNY
TCIAC0721-10G
Cholesterol octyl carbonate
Cholesterol octyl carbonate
分子式: C₃₆H₆₂O₃
MDL编号: MFCD00059873
CAS号: 15455-82-0
https://cn.vwr.com/stibo/web/std.lang.all/77/67/28937767.jpg

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Specific rotation [a]20/D -25.0 to -30.0 deg (C=2,chloroform)
Solubility in Chloroform very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR