4-Amino-3-chlorophenol hydrochloride ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Warning

TCIAA1376-25GEA 995 CNY
TCIAA1376-25G
4-Amino-3-chlorophenol hydrochloride ≥98.0% (通过HPLC,总氮)
4-Amino-3-chlorophenol hydrochloride
分子式: C₆H₇Cl₂NO
MDL编号: MFCD00143110
CAS号: 52671-64-4
https://cn.vwr.com/stibo/web/std.lang.all/93/04/28929304.jpg

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR