2,6-Xylidine hydrochloride ≥98.0% (通过HPLC,总氮)
供应商: TCI EUROPE
TCIAX0029-25GEA
0
CNY
TCIAX0029-25G
2,6-Xylidine hydrochloride ≥98.0% (通过HPLC,总氮)
2,6-Xylidine hydrochloride
分子式:
C₈H₁₂ClN 分子量: 157,6406 g/mol |
MDL编号:
MFCD00060213 CAS号: 21436-98-6 EINECS: 244-388-1 |
产品技术规格
Appearance | White powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...