4-Biphenylboronic acid pinacol ester ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAB3019-5GEA 0 CNY
TCIAB3019-5G
4-Biphenylboronic acid pinacol ester ≥98.0% (通过GC)
4-Biphenylboronic acid pinacol ester
https://cn.vwr.com/stibo/web/std.lang.all/44/62/28934462.jpg

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR