4-硝基-2,6-二苯基苯酚 ≥98.0% (通过滴定分析)
供应商: TCI EUROPE产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...