四氢噻喃-4-酮 99%
供应商: Thermo Scientific
|
Warning
|
ACRO167980050EA
0
CNY
ACRO167980050
四氢噻喃-4-酮 99%
四氢噻喃-4-酮
分子式:
C₅H₈OS 分子量: 116,18 g/mol 溶点: 62…65 °C 存储条件: Ambient |
MDL编号:
MFCD00006660 CAS号: 1072-72-6 EINECS: 214-015-7 |
产品技术规格
Appearance | White to light yellow Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 59 to 63 °C |
GC | ≥98.5 % |
Calcium (Ca) | ≤20 ppm |
Copper (Cu) | ≤10 ppm |
Iron (Fe) | ≤20 ppm |
Lead (Pb) | ≤10 ppm |
Magnesium (Mg) | ≤10 ppm |
Nitrogen (N) | ≤20 ppm |
Phosphorus (P) | ≤50 ppm |
Potassium (K) | ≤20 ppm |
Sodium (Na) | ≤50 ppm |
Zinc (Zn) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...