2'-羟基-4',5'-二甲氧基苯乙酮 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAH1206-5GEA
0
CNY
TCIAH1206-5G
2'-羟基-4',5'-二甲氧基苯乙酮 ≥98.0% (通过GC)
2'-羟基-4',5'-二甲氧基苯乙酮
分子式:
C₁₀H₁₂O₄ 分子量: 196,1999 g/mol |
MDL编号:
MFCD00017241 CAS号: 20628-06-2 |
产品技术规格
Purity (GC) | min. 98.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...