1-甲基-2-吡咯烷酮

供应商: LOCAL DEALS CHINA
Danger

Synonyms: NMPN-甲基-2-吡咯烷酮N-甲基吡咯烷酮

CHLCR01134410100EA 0 CNY
CHLCR01134410100
1-甲基-2-吡咯烷酮
1-甲基-2-吡咯烷酮
分子式: C₅H₉NO
分子量: 99,13 g/mol
沸点: 202 °C (1013 hPa)
溶点: –24 °C
密度: 1,033 g/cm³ (20 °C)
闪点: 91 °C
存储条件: Ambient
MDL编号: MFCD00003193
CAS号: 872-50-4
EINECS: 212-828-1
Merck Index: 12,06197
https://cn.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR