Ethanol ≥75% (v / v)

供应商: LOCAL DEALS CHINA
Danger

CHLCR02544750012EA 0 CNY
CHLCR02544750012
Ethanol ≥75% (v / v)
乙醇
分子式: C₂H₅OH
分子量: 46,07 g/mol
存储条件: Ambient
MDL编号: MFCD00003568
CAS号: 64-17-5
EINECS: 200-578-6
UN号: 1170
ADR: 3,III

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR