2-丙醇

供应商: Merck

Danger

Synonyms: 仲丙醇异丙基醇IPA异丙醇

MERC60104025001730EA 0 CNY
MERC60104025001730
2-丙醇
2-丙醇
分子式: (CH₃)₂CHOH
分子量: 60,1 g/mol
沸点: 82 °C (1013 hPa)
溶点: –89 °C
密度: 0,786 g/cm³ (20 °C)
闪点: 12 °C
存储条件: Ambient
MDL编号: MFCD00011674
CAS号: 67-63-0
EINECS: 200-661-7
UN号: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://cn.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR