Slot blot manifold, PR648
供应商: Cytiva
GEHEPR648EA 0 CNY
GEHEPR648
Slot blot manifold, PR648
PR 648 slot blot filtration manifolds are designed for ease of use and maximum filtration with minimum chance of cross contamination. They make screening of many samples quick and easy.
  • Filter DNA, RNA or protein into clearly defined slots on nitrocellulose or nylon membranes
  • Clamp membranes securely with raised well edges to eliminate cross-contamination without O-rings or gaskets
  • Transfer samples from microplates to wells with standard multichannel pipettors
  • Track sample locations with numbered slots and notched membrane shape
PR 648 slot blot filtration manifolds are designed for reproducibility. Large screws and rim design produce well defined slots and prevent sample leakage between wells. Up to 1 ml can be filtered onto the 6,0×0,8 mm slot area.