illustra™ MicroSpin™ S-200 HR columns
供应商: Cytiva
GEHE27-5120-01EA 0 CNY
GEHE27-5120-01
illustra™ MicroSpin™ S-200 HR columns
核酸试剂 核酸纯化试剂盒和试剂
illustra™ MicroSpin™ S-200 HR columns allow rapid purification of labelled single-stranded or double-stranded DNA fragments >50 bases in length in a volume of 51 to 100 μl.