Cation exchange chromatography column, Mono S HR
供应商: Cytiva
GEHE17-0507-01EA 0 CNY
GEHE17-0507-01
Cation exchange chromatography column, Mono S HR
色谱柱
Mono S HR is a strong cation exchanger prepacked with MonoBeads™. The column is an excellent choice for the polishing step in purification of proteins, peptides, and other biomolecules when high purity is required.
  • Monodisperse 10 µm porous beads yield high resolution, dynamic capacity, reproducibility, and durability
  • Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q ion exchangers
  • Compatible with ÄKTA design, FPLC System, and other high-performance LC systems
Not suitable for use with ÄKTAprime plus system.