Gene Pulser®/Micropulser™ electroporation cuvettes are high-quality electroporation cuvettes which enable reproducible and consistent gene delivery to valuable transfection samples.
- Universal electroporation systems
- Wash thoroughly to eliminate trace inhibitor materials
- Sterilised and individually wrapped
- Reliable sturdy, seamless construction with gap widths set to exacting tolerances for consistent performance
Multiple configurations available in three gap widths and sample type: 0,4, 0,2, and 0,1 cm to deliver the proper field strength to a variety of cell types:
0,1 cm (brown cap) - E. coli, bacteria, yeast
0,2 cm (green cap) - E. coli, bacteria, yeast, mammalian cells, including primary and stem cells
0,4 cm (blue cap) - Mammalian cells, including primary and stem cells, and plant protoplasts