γ-Methacryloxypropyltrimethoxysilane, PlusOne™ Bind-Silane
GEHE17-1330-01EA 0 CNY
GEHE17-1330-01
γ-Methacryloxypropyltrimethoxysilane, PlusOne™ Bind-Silane
PlusOne™ Bind-Silane (γ-methacryloxypropyltrimethoxysilane) is used to covalently attach polyacrylamide gels to a glass surface during staining and drying procedures.
  • Soluble in methanol, diethyl ether and acetone
Used in IEF, PAGE and Sequencing