Pipetting package with macro controller, BLAUBRAND®
供应商: Brand
BRND26007EA 855.27 CNY
BRND26008 BRND26006 BRND26007 BRND26010
Pipetting package with macro controller, BLAUBRAND®
Each pipetting package contains:
  • One macro pipette controller, grey (0,1 - 200 ml)
  • Six BLAUBRAND® graduated pipettes (Type 2), three pipettes 5 ml and 10 ml each, class AS, DE-M marking, with batch certificate
  • Useful product information: BLAUBRAND® volumetric instruments, handling of pipettes (either in German, English, French, Italian or Spanish)
  • Handy plastic container: Ideal for storage of pipettes up to 360 mm length