CoolCell® LX and 5 ml LX cell freezing containers provide consistent, controlled-rate, highly reproducible -1 °C per minute alcohol-free cell freezing when used in a -80 °C freezer. With the elimination of alcohol, freezing results are highly reproducible. CoolCell® containers are unbreakable, open easily when frozen, and are not cold to the touch when frozen. Enhanced design features include beveled junction between base and lid to allow quicker opening once frozen, numbered vial wells for easy sample identification, and exposed vial tops when container is open allowing for faster removal of vials for placement onto dry ice or liquid nitrogen.
- No pre-cooling required
- No waiting between freeze cycles
- -1 °C/minute freeze rate for all cells
- No on-going cost or maintenance
CoolCell® LX holds 12 each 1 or 2 ml vials and CoolCell® 5 ml LX holds 12 each 3,5 to 5 ml vials. The unique materials and solid core design ensures no variability and equal or better post-thaw cell viability than alcohol-based cell freezing containers.