p-Acetoacetophenetidide ≥98.0% (通过GC,总氮)

供应商: TCI EUROPE
Danger

TCIAA2039-25GEA 360 CNY
TCIAA2039-25G
p-Acetoacetophenetidide ≥98.0% (通过GC,总氮)
p-Acetoacetophenetidide
分子式: C₁₂H₁₅NO₃
分子量: 221,2524 g/mol
MDL编号: MFCD00043937
CAS号: 122-82-7
EINECS: 204-577-1
UN号: 2811
ADR: 6.1,III
https://cn.vwr.com/stibo/web/std.lang.all/01/83/28930183.jpg

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR