4,4'-Bis(methylamino)benzop ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
TCIAB1433-1GEA
0
CNY
TCIAB1433-1G
4,4'-Bis(methylamino)benzop ≥98.0% (通过滴定分析)
4,4'-Bis(methylamino)benzop
分子式:
C₁₅H₁₆N₂O |
MDL编号:
MFCD00143227 CAS号: 3708-39-2 |
产品技术规格
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 132.0 to 135.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...