3,4-二甲基二苯甲酮 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD3289-25GEA 0 CNY
TCIAD3289-25G
3,4-二甲基二苯甲酮 ≥98.0% (通过GC)
3,4-二甲基二苯甲酮
分子式: C₁₅H₁₄O
分子量: 210,28 g/mol
沸点: 335°C
MDL编号: MFCD00008525
CAS号: 2571-39-3
EINECS: 219-916-9
https://cn.vwr.com/stibo/web/std.lang.all/02/64/28980264.jpg

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 and le 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR