10,12-Nonacosadiynoic acid ≥97.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAN0514-100MGEA
0
CNY
TCIAN0514-100MG
10,12-Nonacosadiynoic acid ≥97.0% (通过滴定分析)
10,12-Nonacosadiynoic acid
分子式:
C₂₉H₅₀O₂ 存储条件: Refrigerator |
MDL编号:
MFCD00060119 CAS号: 66990-34-9 |
产品技术规格
Appearance | White to Gray to Dark blue powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...