2,4-Heneicosadiynoic acid ≥95.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAH0668-100MGEA
0
CNY
TCIAH0668-100MG
2,4-Heneicosadiynoic acid ≥95.0% (通过滴定分析)
2,4-Heneicosadiynoic acid
分子式:
C₂₁H₃₄O₂ 存储条件: Freezer |
MDL编号:
MFCD00060117 CAS号: 69288-33-1 |
产品技术规格
Appearance | White to Gray to Dark blue powder to crystal |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 73.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...