2,4-Nonadecadiynoic acid ≥97.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAN0515-100MGEA 0 CNY
TCIAN0515-100MG
2,4-Nonadecadiynoic acid ≥97.0% (通过滴定分析)
2,4-Nonadecadiynoic acid
分子式: C₁₉H₃₀O₂
存储条件: Freezer
MDL编号: MFCD00060116
CAS号: 76709-55-2
https://cn.vwr.com/stibo/web/std.lang.all/96/79/28989679.jpg

现在订购

产品技术规格

Appearance White to Gray to Dark blue powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 67.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR