氯甲酸对甲苯酯 97%

供应商: Thermo Scientific
Danger

ACRO314560050EA 0 CNY
ACRO314560050
氯甲酸对甲苯酯 97%
氯甲酸对甲苯酯
分子式: C₈H₇ClO₂
沸点: 99,5 °C (20 mmHg)
闪点: 86 °C
存储条件: Refrigerator
MDL编号: MFCD00013255
CAS号: 937-62-2
EINECS: 213-332-8
UN号: 3277
ADR: 6.1,II

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5100 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR