3-Nitroaniline hydrochloride ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAN0120-25GEA
0
CNY
TCIAN0120-25G
3-Nitroaniline hydrochloride ≥98.0% (通过HPLC,滴定分析)
3-Nitroaniline hydrochloride
分子式:
C₆H₇ClN₂O₂ 分子量: 174,5900 g/mol |
MDL编号:
MFCD00050693 CAS号: 33240-96-9 EINECS: 251-421-3 ADR: 6.1,II |
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...