4-丙烯氧基-2-羟基二苯甲酮 ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAA2342-25GEA
0
CNY
TCIAA2342-25G
4-丙烯氧基-2-羟基二苯甲酮 ≥98.0% (通过HPLC,滴定分析)
4-丙烯氧基-2-羟基二苯甲酮
分子式:
C₁₆H₁₄O₃ 分子量: 254,2806 g/mol 沸点: 178°C |
MDL编号:
MFCD00191713 CAS号: 2549-87-3 |
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...