4-氟-4'-羟基二苯甲酮 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAF0269-10GEA
0
CNY
TCIAF0269-10G
4-氟-4'-羟基二苯甲酮 ≥98.0% (通过GC)
4-氟-4'-羟基二苯甲酮
分子式:
C₁₃H₉FO₂ 分子量: 216,2078 g/mol |
MDL编号:
MFCD00011484 CAS号: 25913-05-7 EINECS: 247-340-8 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 168.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...