二乙酸三苯铋(V) ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
TCIAT1836-5GEA
0
CNY
TCIAT1836-5G
二乙酸三苯铋(V) ≥98.0% (通过滴定分析)
二乙酸三苯铋(V)
分子式:
C₂₂H₂₁BiO₄ |
MDL编号:
MFCD00092697 CAS号: 7239-60-3 |
产品技术规格
Appearance | White to Yellow to Green powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Solubility in hot Toluene | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...